Skip to Content
MilliporeSigma
All Photos(7)

Documents

CWA-1003

Sigma-Aldrich

Anti-Mouse CD23 (2G8) ColorWheel® Dye-Ready mAb

greener alternative

for use with ColorWheel® Dyes (Required, sold separately)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

2G8, monoclonal

product line

ColorWheel®

form

lyophilized

mol wt

calculated mol wt 37.65 kDa

species reactivity

mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable

isotype

IgG2bκ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

compatibility

for use with ColorWheel® Dyes (Required, sold separately)

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

mouse ... Fcer2(14128)

General description

ColorWheel® technology is a novel and proprietary method of creating your own antibody and dye combinations for use in flow cytometry. By incubating any ColorWheel® antibody with any ColorWheel® dye, researchers can quickly and simply produce primary conjugated antibodies for use in single color or multicolor flow cytometry analysis. Each ColorWheel® antibody and dye is lyophilized for long long-term storage, allowing for simplicity and flexibility without compromise. ColorWheel® technology requires both ColorWheel® antibody and ColorWheel® dye for Flow Cytometry application.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This product is Preservative-free, lyophilized product for enhanced stability and allow for ambient shipping and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.

Specificity

Clone 2G8 is a rat monoclonal antibody that detects murine CD23.

Immunogen

FceR isolated from murine B cell lymphoma cell line M12.4.5

Application

Quality Control Testing:

Evaluated by Flow Cytometry in Mouse splenocytes.

Flow Cytometry Analysis: Staining of one million mouse splenocytes was performed using 5 μL of 1:1 mixture of Cat. No. CWA-1003, Anti-Human CD23 (2G8) ColorWheel® Dye-Ready mAb and CWD-PE, ColorWheel® Antibody-Ready Phycoerythrin (PE) Dye or an equivalent amount of a PE-conjugated Rat IgG2a isotype control.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Compatibility

Requires ColorWheel® Dye (Sold Separately)

Target description

Low affinity immunoglobulin epsilon Fc receptor (UniProt: P20693; also known as Fc-epsilon-RII, Lymphocyte IgE receptor, CD23) is encoded by the Fcer2 (also known as Fcr2a) gene (Gene ID: 14128) in murine species. CD23 is a homotrimeric, single-pass type II membrane glycoprotein that contains a cytoplasmic domain (aa 1-23); a transmembrane domain (aa 24-49), and an extracellular domain (aa 50-331). The extracellular domain presents striking similarity to C-type lectins. CD23 expression is reported in B cells, monocyte, follicular dendritic cells, platelets, and eosinophils. It is also expressed in cases of chronic lymphocytic leukemia (CLL) and in follicular lymphomas. It serves as a low-affinity receptor for immunoglobulin E (IgE) and CR2/CD21 and plays an essential role in the regulation of IgE production and in B-cell differentiation. In addition to its role as a receptor for IgE, CD23 can also be released from cell surface as a soluble protein (sCD23) via ADAM10-mediated ectoderm shedding. Mice lacking ADAM10 expression in their B cells display defective release of CD23. Clone 2G8 is reported to interact at a site similar to but not identical with the site on CD23 that binds IgE. (Ref.: Kintner, C., and Sugden, B. (1981). Nature. 294(3); 458-460; Margaret, E., et al. (1999). J. Immunol. 163(1); 217-223; Acharya, M., et al. (2010). Clin. Exp. Immunol. 162(1); 12-23).

Physical form

Lyophilized from PBS containing D-Mannitol and Sucrose, normal appearance is a dried pellet. Reconstituted antibody solution is stable and functional as assessed by functional testing. Contains no biocide or preservatives, such as azide.

Reconstitution

1.0 mg/mL after reconstitution at 25 μL in PBS. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C. Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial. Reconstitute each vial by adding 25 μL of PBS. Protect from light.

Legal Information

ColorWheel is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service