Skip to Content
MilliporeSigma
All Photos(1)

Documents

CRISPR21

Sigma-Aldrich

Lenti CRISPR Human HPRT1 Positive Control Plasmid DNA (LV04 vector)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106610
NACRES:
NA.51

form

liquid

Quality Level

packaging

vial of 50 μL

concentration

20 ng/μL in TE buffer; DNA (1μg of plasmid DNA)

application(s)

CRISPR

shipped in

dry ice

storage temp.

−20°C

General description

The Lenti CRISPR Human HPRT1 Positive Control Plasmid DNA (LV04 vector) is a critical positive control to monitor transfection efficiency for lentiCRISPRs. The protein encoded by this gene is a transferase which plays a central role in the generation of purine nucleotides. Cells with intact HPRT1 will die upon exposure to 6-TG (Product No. A4882), an antimetabolite used in the treatment of leukemias. Conversely, cells with HPRT1 knocked out will increase over time with 6-TG selection. This targets a validated HPRT1 CRISPR site, which serves as an experimental control for lenti CRISPRs. The lenti CRISPR positive control is a dual-component system consisting of a U6-driven guide RNA plasmid targeting the human HPRT1 gene and hPGK-driven Puromycin and BFP expression. This control requires Cas9 be delivered by a separate vector.

Application

Functional Genomics/Screening/Target Validation

Other Notes

Ampicillin and puromycin antibiotic resistance genes provide selection in bacterial or mammalian cells, respectively. Self-inactivating replication incompetent viral particles can be produced in packaging cells (HEK293T) by co-transfection with compatible packaging plasmids (Product No. SHP002).

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Genome-wide screening with optimized gRNAs per gene ensures specific and efficient knockout, controlling time and cost.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service