Skip to Content
MilliporeSigma
All Photos(2)

Documents

CN304-05

Sigma-Aldrich

Canine Aortic Endothelial Cells: CnAOEC (Cryovial)

Synonym(s):

Endothelial cell line

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

biological source

canine aorta

packaging

pkg of 500,000 cells

manufacturer/tradename

Cell Applications, Inc

growth mode

Adherent

karyotype

2n = 78

morphology

Endothelial

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

diabetes; cardiovascular diseases

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.


CnAOEC provide a useful model to study cardiovascular diseases and test potential therapeutic agents, not only important from a veterinary perspective, but also having implications for human health. In a study aiming to adapt an immunomagnetic isolation protocol used in human patients to isolate circulating endothelial cells in dogs, CnAOEC cells were shown to be positive for von Willebrand factor, CD146 and stained with Ulex europaeus agglutinin 1 (Wills, 2009). To investigate the mechanism of N-terminal portion of pro C-type natriuretic peptide (NT-pCNP), a sepsis biomarker in humans and dogs, CnAOEC were treated with a variety of agents and it was found that lipopolysaccharide, TNF-α and IL-1β, but not IL-6, IL-10, IL-21, CXCL-8, IFN-γ, VEGF-A, stimulated NT-pCNP production (Osterbur, 2012, 2013). CnAOEC were also used to demonstrate vasoprotective activity of pomegranate and soy isoflavons in order to prevent endothelial disfunction in dogs which commonly leads to cardiovascular disease (Baumgartner-Parzer, 2012). Finally, CnAOEC were utilized as a control to compare the growth of normal canine endothelial cells and aggressive canine hemangiosarcoma cells in a study investigating signaling pathways underlying hemangiosarcoma oncogenesis (Murai, 2012), and as a VEGFR2-positive control in a study aimed at understanding the reasons for reduced numbers of circulating progenitor cells in cardiovascular disease (Boilson, 2010).

Cell Line Origin

Aorta

Application

cardiovascular function, studies on immune system and graft rejection, development of 3D endothelialized engineered tissues, drug discovery, stent-graft compatibility testing

Components

Canine Endothelial Cell Basal Medium containing 10% FBS & 10% DMSO

Preparation Note

  • 2nd passage, >500,000 cells in Canine Endothelial Cell Basal Medium containing 10% FBS & 10% DMSO
  • Can be cultured at least 16 doublings

Subculture Routine

Please refer to the CnAOEC Culture Protocol.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service