Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

CMR0001

Sigma-Aldrich

Recovery Medium for Expression

for use with competent cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.85

grade

Molecular Biology

sterility

sterile; autoclaved

shipped in

dry ice

storage temp.

−20°C

General description

This recovery media has and optimized formulation for ensuring stability and high-efficiency transformation of protein expression clones.

Features and Benefits

  • Formulated without lactose to prevent unwanted leaky expression from T7 or other lactose/IPTG-inducible promoters.
  • Enables the highest possible transformation efficiency.
  • Use in the outgrowth stage of competent cell transformation to achieve 10-folder higher transformation efficiency than you would get with LB or SOC media.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Salvatore Di Girolamo et al.
Microbial cell factories, 19(1), 170-170 (2020-08-29)
Miniaturization of biochemical reaction volumes within artificial microcompartments has been the key driver for directed evolution of several catalysts in the past two decades. Typically, single cells are co-compartmentalized within water-in-oil emulsion droplets with a fluorogenic substrate whose conversion allows

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service