Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

CMC0003

Sigma-Aldrich

SIG10 HIGH Electrocompetent Cells

for protein expression and DNA plasmid production

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.85

biological source

Escherichia coli

grade

for molecular biology

form

buffered aqueous solution

growth mode

adherent or suspension

morphology

rod shaped

technique(s)

microbiological culture: suitable

cell transformation

competent cell type: electrocompetent
transformation efficiency: ≥5 × 109 cfu/μg

shipped in

dry ice

storage temp.

−70°C

General description

The SIG10 HIGH Electrocompetent Cells are high efficiency cells with a substantially lower cost per reaction and are the most economical choice for standard cloning and library construction.

Genotype

F- mcrA Δ(mrr-hsdRMS-mcrBC) endA1 recA1 Φ80dlacZΔM15 ΔlacX74 araD139 Δ(ara,leu)7697galU galK rpsL nupG λ- tonA (StrR)

Features and Benefits

  • share the most useful genetic elements of standard cloning strains like DH5a, DH10B, JM109, TOP10, etc. and directly replace them in cloning protocols.
  • incorporate a unique manufacturing technology that increases transformation efficiency, recombinant yields and reliability.
  • provide solutions for a wide range of applications at economical prices.
  • Blue - white screening

Components

  • SIG10 HIGH electrocompetent cells
  • pUC 19 transformation control DNA
  • recovery medium for cloning

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service