Key Documents
CLS766119
Pyrex® pennyhead stopper
stopper joint: 19
Synonym(s):
glass stopper, glass stoppers, stoppers, Corning® 7660
Select a Size
6 EA
$166.00
$166.00
Please contact Customer Service for Availability
Select a Size
Change View
6 EA
$166.00
About This Item
UNSPSC Code:
41122500
NACRES:
NB.43
$166.00
Please contact Customer Service for Availability
material
glass
Joint
joint: stopper19
manufacturer/tradename
Corning 7661-19
General description
Ground zone length indicates its greatest diameter (mm).
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)