CLS6798610D
Corning® stirrer with digital display
AC/DC input 230 V AC (CEE7-7 plug), plate L × W 10 in. × 10 in.
Synonym(s):
magnetic stirrer, magnetic stirrers, stirrers
Select a Size
$1,060.00
Select a Size
About This Item
$1,060.00
Recommended Products
packaging
case of 1
manufacturer/tradename
Corning 6798-610D
availability
not available in USA, Canada and EU
parameter
60-1150 rpm speed
plate L × W
10 in. × 10 in.
General description
- Digital LED stir speed display indicates when set speed has been reached
- Microprocessor maintains consistent and repeatable stir speed settings
- Extremely durable and heat resistant Pyroceram glass-ceramic top
- Small footprint and low profile
- Meets UL and cUL standards
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)