Key Documents
CLS6705
Corning® LSE Platform Rocker
Double PlatformDouble Platform, AC/DC input 115 V, AC/DC input 50/60 Hz, AC/DC input, US 3-pin plug
Synonym(s):
Laboratory rocker, Platform shaker, Rocking platform
Select a Size
1 EA
$1,620.00
$1,620.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
$1,620.00
About This Item
UNSPSC Code:
41103811
$1,620.00
Please contact Customer Service for Availability
feature
Double Platform
Double Platform
manufacturer/tradename
Corning 6705
6705
General description
The Corning LSE Platform Rocker is a compact yet high capacity rocker that is ideal for western blotting and general mixing applications. The adjustable rocking speed and fixed 7 degree tilt makes it optimal for staining fragile gels, as well as the more intensive action necessary for washing blots.
The speed of the Corning Platform Rocker can be adjusted from 2 to 60 rpm and the timer can be set for up to 2 hours or for continuous operation. The unit is safe to use in temperature controlled environments up to 70°C and is available in single or high capacity double platform configurations.
Features and Benefits
  • Low profile and compact size
  • 11-pound load capacity
  • Non-slip rubber pads to prevent sample containers from moving during operation
  • Double platform model available for increased capacity
Legal Information
Corning is a registered trademark of Corning, Inc.
LSE is a trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)