Key Documents
CLS6406125FO
PYREX® replacement Squibb separatory funnel w/o valve and stopper
Synonym(s):
equipment replacement accessory
Select a Size
4 EA
$425.00
$425.00
Please contact Customer Service for Availability
Select a Size
Change View
4 EA
$425.00
About This Item
UNSPSC Code:
41121809
$425.00
Please contact Customer Service for Availability
packaging
pack of 4 ea
manufacturer/tradename
Corning 6406-125FO
capacity
125 mL
volume
125 mL
Related Categories
General description
These economy Squibb separatory funnel replacements do not have a Polytetrafluorethylene (PTFE) valve assembly or polyethylene stopper.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)