Key Documents
CLS4923
Corning® pipetting aids
Air filter replacement
About This Item
UNSPSC Code:
41121500
Pricing and availability is not currently available.
manufacturer/tradename
Corning 4923
General description
Costar Stripettors are lightweight, adjustable-speed pipetting aids designed for use with all serological pipettes. The nose cone of the units are autoclavable and fitted with a replaceable 0.2 μm hydrophobic sterilizing filter for added protection against biohazardous material. The Stripettor operates on a rechargeable 9V NiCd battery and features an LED light on the handle that lets the user know when to recharge. The unit is fully operational while recharging.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)