Skip to Content
MilliporeSigma
All Photos(1)

Documents

CLS4826

Corning® microvolume pipet tips

0.1-10 μL, universal fit racked pipet tips, natural, non-sterile, 96 tips/rack, 960tips/case

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.01

material

natural tip
polypropylene

sterility

non-sterile

feature

DNase free
RNase free
barrier: no
disposable
endotoxin free
filter: no
graduation

packaging

rack of 96

manufacturer/tradename

Corning 4826

volume range

0.1-10 μL

compatibility

for use with Gilson® and other popular ultra-micropipettors

Looking for similar products? Visit Product Comparison Guide

General description

Microvolume Tips provide accurate, reliable performanace for major brand ultra-pipettors. All racked tips are certified RNase/DNase-free and nonpyrogenic.

Legal Information

Corning is a registered trademark of Corning, Inc.
Gilson is a registered trademark of Gilson, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service