Skip to Content
MilliporeSigma
All Photos(1)

Documents

CLS4513

Corning® 384 well microplate, low volume

surface treatment, Non-binding Surface, white polystyrene, U-bottom, non-sterile, lid: no, U-bottom (white), pack of 10

Synonym(s):

Corning® 384 well microplate, low volume | Sigma-Aldrich

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.24

material

U-bottom
white bottom
white polystyrene

sterility

non-sterile

feature

lid: no
skirt
U-bottom (white)

packaging

pack of 10

manufacturer/tradename

Corning 4513

well volume

35 μL

wells

384 wells , low volume

working volume

2-20 μL

binding type

NBS (non-binding surface)

Looking for similar products? Visit Product Comparison Guide

General description

The Corning® 384-well microplate, low volume features a skirted, U-bottom design with white walls, made from white polystyrene material, that contributes to enhanced luminescent signals by reducing well-to-well crosstalk and minimizing background luminescence and fluorescence. Additionally, incorporation of the nonbinding surface (NBS) technology helps create a nonionic hydrophilic surface similar to polyethylene oxide, which effectively reduces protein and nucleic acid binding at low concentrations.

Application

Corning®384 well microplate is suitable for the assay signal-to-noise ratio.

Features and Benefits

  • It has a square well top that facilitates robotic pipetting
  • Rounded bottom to eliminate trapped air and boost the Z factor
  • Light cone-shaped conical well for better efficiency
  • White walls reduced fluorescence and crosstalk between wells during fluorescent experiments
  • Microplates have non-ionic hydrophilic surfaces due to nonbinding surface (NBS) technology treatment that reduces molecular interactions
  • Raised rim for better sealing

Legal Information

Corning is a registered trademark of Corning, Inc.
NBS is a trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service