CLS440826
Corning® nine-position stirrer integral control
AC/DC input 120 V AC, US 3-pin plug
Synonym(s):
Corning stirrer accessory
Select a Size
$2,910.00
Select a Size
About This Item
$2,910.00
Recommended Products
manufacturer/tradename
Corning 440826
General description
- Compact design saves valuable bench space
- Offers powerful stirring from 350 to 2000 rpm.
- Rugged lightweight glass filled resin case
- Two-year warranty
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)