CLS431227
Corning® syringe filters
PTFE membrane, diam. 50 mm, pore size 0.2 μm
Synonym(s):
disposable syringe filters, sterile syringe filters, syringe filters
Select a Size
$244.00
Select a Size
About This Item
$244.00
Recommended Products
material
PTFE membrane
polypropylene housing
sterility
sterile
packaging
case of 12
manufacturer/tradename
Corning 431227
diam.
50 mm
pore size
0.2 μm
fitting
hose barb
Related Categories
General description
PES for low protein binding and faster flow rates; surfactant-free cellulose acetate (SFCA) for the lowest protein binding; PTFE for chemical resistance; and regenerated cellulose (RC) the best choice for DMSO compatibility.
- PTFE membranes provide the best chemical resistance
- These naturally hydrophobic membranes can be used for venting applications or filtering gases
- 100% integrity tested, sterile and certified nonpyrogenic and noncytotoxic
- Manufactured in accordance with ISO 9002 standards
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)