Documents
CLS430180
Corning® polypropylene beaker
sterile, size 250 mL , beaker, container only, case of 500
Synonym(s):
plastic beakers, polypropylene beakers, storage containers
About This Item
UNSPSC Code:
41121803
NACRES:
NB.13
material
natural polypropylene (beaker)
orange high-density polyethylene (snap cap)
sterility
sterile
sterile
feature
container only
packaging
pack of 20
case of 500
manufacturer/tradename
Corning 430180
size
250 mL , beaker
General description
Corning flexible polypropylene container with snap-on polyethylene lid serves as a beaker or storage container.
  • Container is graduated in both millilitres and ounces
  • Certified nonpyrogenic
Other Notes
Browse all plastic beakers
Legal Information
Corning is a registered trademark of Corning, Inc.