Skip to Content
MilliporeSigma
All Photos(1)

Documents

CLS3727BC

Corning® 1536 well plate, with generic barcode

size 1536 wells, flat bottom white polystyrene wells, sterile

Synonym(s):

1536-Well Plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13

material

flat bottom white polystyrene wells
polystyrene plate

sterility

sterile

feature

lid
skirt

packaging

pack of 10
case of 50 ea
case of 50

manufacturer/tradename

Corning 3727BC

maximum volume

12.5 μL

size

1536 wells

working volume

8 μL

color

white

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Bar Code Details

Corning barcode labels have been quality tested for optimal readability, chemical resistance and temperature variation.

  • Resistant to commonly used laboratory reagents (100% DMSO, Methanol, Ethanol, Ethyl Acetate, Methylene Chloride, Isopropanol, Acetronitrile, Ethanol/Water/TFA, Sodium Hydroxide, Hydrochloric Acid)
  • Resistant to temperature ranges of -80ºC to +52ºC

Technical Details
  • Bar Code labeling specifications
  • 2.75" x 0.25" label size
  • Code 128
  • Code 3 of 9
  • 10 mil narrow bar element (x-dimension = 0.010")

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service