Skip to Content
MilliporeSigma
All Photos(1)

Documents

CLS3570BC

Corning® 384 well microplate, low volume

Tissue Culture (TC)-treated surface, white polystyrene, flat bottom white, sterile, with generic barcode, lid, pack of 10

Synonym(s):

384 multiwell plate, 384 well barcode plate, 384 well microplate, 384 well microtiter plate, 384 well plate, 384 well plate with generic barcode

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.24

material

flat bottom white
square white wells
white bottom
white polystyrene

sterility

sterile

feature

lid
skirt
with generic barcode

packaging

pack of 10

manufacturer/tradename

Corning 3570BC

well volume

112 μL

wells

384 wells

working volume

20-80 μL

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service