Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS356516

Corning® BioCoat Poly-L-Lysine Microplate

well size 96 wells, flat bottom clear wells, lid

Synonym(s):

Cell Culture Plate, plastic coated cell culture plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122102
NACRES:
NB.13

material

flat bottom clear wells

feature

lid

packaging

pkg of 50 ea (10 packs of 5)

manufacturer/tradename

Corning 356516

well size

96 wells

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

Application

Cell culture

Legal Information

BioCoat is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service