Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

CLS353935

Corning® Falcon® Cell Culture Plate

wells, 24, Tissue Culture (TC)-treated surface, flat bottom clear wells, sterile

Synonym(s):

Cell Culture Plate

Sign Into View Organizational & Contract Pricing

Select a Size

60 EACH
$303.00

$303.00


Please contact Customer Service for Availability


Select a Size

Change View
60 EACH
$303.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
flat bottom clear wells
size:
24 well
sterility:
sterile
binding type:
Tissue Culture (TC)-treated surface
feature:
lid
wells 24

$303.00


Please contact Customer Service for Availability

material

flat bottom clear wells

sterility

sterile

feature

lid
wells 24

packaging

case of 60 ea (6 packs of 10)

manufacturer/tradename

Corning 353935

size

24 well

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Falcon® 24 Well Clear Flat Bottom, Tissue Culture-Treated Multiwell Cell Culture Plate with Lid, Sterile, Ready-Stack Tray, 10/Pack, 60/Case

Application

Cell Culture

Legal Information

Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service