CLS353043
Corning® Falcon® Cell Culture Plate
wells, 12, Tissue Culture (TC)-treated surface, flat bottom clear wells, sterile
Synonym(s):
Cell Culture Plate
Select a Size
$234.00
Select a Size
About This Item
$234.00
Recommended Products
material
flat bottom clear wells
sterility
sterile
feature
lid
wells 12
packaging
case of 50 ea (Individually wrapped)
manufacturer/tradename
Corning 353043
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
Application
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service