CLS352095
Corning® Falcon® Centrifuge Tubes
capacity 15 mL, conical polystyrene, cap, dome seal screw, bagged, sterile
Synonym(s):
culture tube
Select a Size
$389.00
Select a Size
About This Item
$389.00
Recommended Products
material
conical polystyrene
sterility
sterile
feature
cap dome seal screw
manufacturer/tradename
Corning 352095
capacity
15 mL
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service