CLS351058
Corning® Falcon® Bacteriological Petri Dish
W × H 150 mm × 15 mm, lid, standard, sterile
Synonym(s):
plastic cell culture dish
Select a Size
$212.00
Select a Size
About This Item
$212.00
Recommended Products
material
polystyrene
sterility
sterile
feature
lid standard
manufacturer/tradename
Corning 351058
technique(s)
microbiological culture: suitable
W × H
150 mm × 15 mm
Looking for similar products? Visit Product Comparison Guide
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service