Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS3346

Corning ® 96 Well Microplate Storage Mat

non-sterile

Synonym(s):

96 multiwell plate

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
$254.00
50 EA
$977.00

$254.00


Please contact Customer Service for Availability


Select a Size

Change View
10 EA
$254.00
50 EA
$977.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
size:
96 wells

$254.00


Please contact Customer Service for Availability

description

for expanded volume 450 μL plates
storage mat

sterility

non-sterile
non-sterile

manufacturer/tradename

Corning 3346

size

96 wells

suitability

suitable for (general storage, liquid handling; automated applications)

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service