Skip to Content
MilliporeSigma
All Photos(1)

Documents

CLS3345

Corning® microplate sealing tape

white Rayon film (with acrylic), sterile, suitable for (for cell/tissue culture applications, breathable sterile membrane)

Synonym(s):

microplate sealing film, multiwell plate sealing film, sealing film

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122410
NACRES:
NB.15

material

white Rayon film (with acrylic)

sterility

sterile

packaging

case of 500 ea

manufacturer/tradename

Corning 3345

W × L

3.25 in. × 6.00 in.
8.26 cm × 15.24 cm

color

clear

suitability

suitable for (for cell/tissue culture applications, breathable sterile membrane)

Looking for similar products? Visit Product Comparison Guide

General description

  • Seals polypropylene and polystyrene culture plates, 96-well plates, 384-well plates and other assay plates
  • Porous adhesive film specially designed for use in cell and tissue culture applications
  • The film seals all types of microplates and effectively allow gas exhcnage for cellular and bacterial cultivation, while preventing cross-contamination
  • Temp. range. -20°C to +80°C
  • Sealing tapes are not autoclavable

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service