Skip to Content
MilliporeSigma
All Photos(1)

Documents

CLS3341

Corning® sealing mat for 384 well deep well plate

white polystyrene, V-bottom, non-sterile, pack of 10

Synonym(s):

384 well deep well plate sealing mat, 384 well plate sealing mat, deep well plate sealing mat, microplate sealing mat, multiwell plate sealing mat, plate sealing mat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13

material

V-bottom
natural wells
white polystyrene

sterility

non-sterile

feature

plate format: 384 well sealing mat
sealing mat

packaging

pack of 10
case of 50

manufacturer/tradename

Corning 3341

size

384 wells

well volume

240 μL

color

natural

General description

  • Precise fit for 384 well deep well polypropylene storage block (CLS3342; CLS3347)
  • Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g. DMSO, ethanol, methanol)
  • Firm seal prevents evaporation and gas exchange
  • Ideal for use in long-term, low temperature compound storage

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service