Skip to Content
MilliporeSigma
All Photos(1)

Documents

CLS3271

Corning® CellSTACK® cell culture chambers

Tissue Culture (TC)-treated surface, clear polystyrene, sterile, graduations, high-density polyethylene cap, case of 6

Synonym(s):

cell culture flasks, scale up cell culture flasks, tissue culture flasks

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122104
NACRES:
NB.13

material

clear polystyrene
high-density polyethylene cap
rectangular bottom flask
short neck

sterility

sterile

feature

cap vented
graduations

packaging

pack of 1
case of 6

manufacturer/tradename

Corning 3271

surface area

6360 cm2

working volume

1250-1900 mL

suitability

suitable for (scale-up cell culture)

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Ideal for cell culture scale-up
  • Culture chambers produce cell yields in the 107 - 109 range
  • Useful alternatives to multiple roller bottles or spinner flasks.
  • High mechanical strength and structural integrity
  • 100% leak tested prior to shipping
  • Certified nonpyrogenic and sterilized by gamma irradiation, with low particulate contamination from modern manufacturing methods
  • Easy to use larger openings with threaded closures and vented caps
  • Large 26 mm wide filling ports allow direct access to chamber bottom, with more options for sterile transfer of media and cultures

Features and Benefits

  • Has 6,360cm2 cell growth area
  • Two 26mm diameter filling ports allow direct access to chamber bottom providing greater flexibility for sterile filling and emptying by pouring, pipetting or via tubing in a fully closed system
  • Standard 33mm threaded caps have 0.2μm pore nonwettable membranes sealed directly to the caps to allow gas exchange while minimizing the risk of contamination
  • Optional 33mm threaded caps are available with integrally sealed USP Class VI certified C-FLEX tubing to allow direct sterile transfer of media and cells
  • USP Class VI polystyrene provides excellent optical clarity and mechanical strength
  • Sterilized by gamma radiation and certifiied nonpyrogenic
  • 100% leak tested prior to shipping

Legal Information

CellSTACK is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service