Key Documents
CLS3087
Corning® multiwell plates, plate lids and sealing mats
Thermowell GOLD 96 well sealing mat, rubber silicone septa mat, pierceable, non-sterile, 50/cs
Synonym(s):
96 well sealing mat, rubber silicone sealing mat, rubber silicone septa sealing mat
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
material:
silicone
material
silicone
sterility
non-sterile
packaging
pack of 10
case of 50
manufacturer/tradename
Corning 3087
General description
  • Offers a unique piercible surface for easy access to reaction contents
  • Compatible with all Thermowell GOLD 96 well PCR microplates
  • Fully autoclavable and reusable for a more cost effective alternative to other sealing methods
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)