CLS3073
Corning® CellBIND® Surface cell culture flasks
CellBIND 100cm2 canted neck low profile flask w/ Vent Cap, PS, sterile, 60/cs
Synonym(s):
cell culture flasks, tissue culture flasks
About This Item
Recommended Products
material
angled neck flask (low profile)
cap (vented)
high-density polyethylene cap
rectangular bottom flask
rectangular flask
rectangular polystyrene
vent cap
sterility
sterile; γ-irradiated
feature
cap
graduations
packaging
pack of 6
case of 60
manufacturer/tradename
Corning 3073
maximum volume
30 mL
surface area
100 cm2
working volume
20-30 mL
suitability
suitable for (cell culture applications; optimal for adherent cell culturing)
binding type
CellBIND® surface
General description
CellBIND may provide a more ecominic alternative to expensive and unstable biological coatings.
Both enzymatic and non-enzymatic dissociating solutions have been used to remove and harvest cells from CellBIND surfaces, scraping is not recommended due to the increased strength of cell attachment.
- Manufactured from optically clear virgin polystyrene.
- Treated CellBIND surface treatment for enhanced cell attachment
- 100% integrity tested.
- Sterilized by gamma irradiation and certified nonpyrogenic