Key Documents
CLS2679940
Corning® polypropylene closure
fits neck O.D. 40 mm
, orange
Select a Size
100 EA
$428.00
$428.00
Please contact Customer Service for Availability
Select a Size
Change View
100 EA
$428.00
About This Item
UNSPSC Code:
41121800
$428.00
Please contact Customer Service for Availability
material
orange
polypropylene
packaging
case of 100
General description
For culture tubes and flasks with long rimless necks. Position closure open for gas exhange or closed for humidified environment. Internal drip ring minimizes contamination.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)