Key Documents
CLS139532HTC
Corning® high-temp PBT screw cap
For high temps, GL 32 solid top, with PTFE faced silicone liner
Select a Size
10 EA
$137.00
$137.00
Please contact Customer Service for Availability
Select a Size
Change View
10 EA
$137.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.11
$137.00
Please contact Customer Service for Availability
material
PTFE liner
red PBT cap
sterility
non-sterile
packaging
pack of 10
fitting
GL 32 solid top
General description
The red Corning polybutylene terephthalate (PBT) high temperature caps have a GL32 thread, can be dry heat sterilized at up to 180 deg. C and have a Polytetrafluorethylene (PTFE) liner for efficient sealing.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)