Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

10T½

99072801, mouse embryo (c3h), Fibroblast-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

10T½, 99072801

biological source

mouse embryo (c3h)

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse embryo fibroblast

Cell Line Description

Clone 8, contact inhibited, semi-normal.

Culture Medium

BME + 10% FBS.

Subculture Routine

Split sub confluent cultures (70-80%) 1:3 ie. seeding at 3x10,000 cells/cm2 using 0.25% trypsin/EDTA. 37°C; 5% CO2

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service