Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

COR-L26

96020740, human lung, Floating aggregates of packed cells. Irregular shaped aggregates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

COR-L26, 96020740

biological source

human lung

growth mode

Suspension

karyotype

Not specified

morphology

Floating aggregates of packed cells. Irregular shaped aggregates

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human large cell lung cancer bone marrow aspirate

Cell Line Description

A human large cell lung cancer bone marrow aspirate derived cell line established from a female patient.

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cell densities between 2 x 100,000 cells/ml and 2 x 1000,000 cells/ml. Saturation density 2 x 1000,000; 37°C; 5% CO2.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service