Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CRE BAG2

95082219, mouse unknown/unspecified, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

CRE BAG2, 95082219

biological source

mouse unknown/unspecified

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse NIH 3T3 fibroblast, Moloney murine leukaemia virus transfected

Cell Line Description

CRE BAG2 has been established from the NIH 3T3 fibroblast cell line. Cells were transfected with Moloney murine leukaemia virus-derived proviral genomes carrying complementary mutations in the gag-pol or env regions. CRE BAG2 produces BAG ( a β-gal-transducing vector) and can be used for the package of vectors derived from murine leukaemia viruses. The production of recombinant replication-competent helper viruses has never been reported.

Application

Virus-packaging assays, cell-lineage marking systems

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:5 to 1:10 i.e. seeding at 1-2x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service