Skip to Content
MilliporeSigma
All Photos(1)

Documents

DMS 153

95062827, human lung (liver metastasis), Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

DMS 153, 95062827

biological source

human lung (liver metastasis)

growth mode

Semi-adherent aggregates

karyotype

Modal no. 41-42

morphology

Not specified

products

Not specified

receptors

Bombesin, Epidermal growth factor (EGF)

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Human Caucasian lung small cell carcinoma, metastasis to liver

Cell Line Description

DMS 153 was isolated from the liver of a 44 year-old male patient at autopsy who had been treated with cytoxan and methotrexate. The cells were identified as small cell carcinoma of the lung (SCLC) and secrete moderate quantities of calcitonin (CT) and calcitonin gene related peptide (CGRP). Production of adrenocorticotropin (ACTH), bombesin (BN), 17 β-estradiol and oxytocin-neurophysin (OT-NP) has also been reported. The expression of cell surface antigens Leu-7, HLA-DR, My 23 (defined by AML-2-23 monoclonal antibody), the AML-1-99 defined antigen and CR3 receptor (defined by the OKM antibody) has been shown. DMS 153 cells produce receptors for BN and epidermal growth factor (EGF) and are tumourigenic in athymic, nude mice.

Application

Human lung tumour model

DNA Profile

STR-PCR Data:
Amelogenin: X,Y
CSF1PO: 10
D13S317: 13
D16S539: 12,13
D5S818: 12
D7S820: 8,10
THO1: 7,9.3
TPOX: 11
vWA: 14,15

Culture Medium

Waymouth′s MB 752/1 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS) (Heat Inactivated).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.Cells grow in large clumps.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service