Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

IMT 9

95060901, deer skin, Fibroblast-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

IMT 9, 95060901

biological source

deer skin

growth mode

Adherent

karyotype

Stable 2n = 6 genome

morphology

Fibroblast-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Indian Muntjac skin fibroblast

Cell Line Description

The cell line IMT 9 has been derived from the non-transformed skin fibroblast cell line Indian Muntjac (Sigma product no. 90112813). Transformation was achieved by continuous exposure to low-level cadmium (5 μM) and a short non-toxic single dose of methyl-nitroso urethane (MNU, 24 h at 0.1μm). Exposure to cadmium led to the establishment of SCR5 (ECACC product no. 95071120); the MNU treatment of SCR5 cells followed by tumour formation in a nude mouse gave rise to the cell line IMT9. IMT9 is highly tumourigenic whereas SCR5 expresses a low tumour level. The cells have been eradicated from mycoplasma at ECACC.

Culture Medium

MEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:8 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.
Depositor and originator: Dr. M. Ord, University of Southampton, Department of Biology, School of Biological Sciences, Southampton, UK.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service