Skip to Content
MilliporeSigma
All Photos(1)

Documents

NCI-H929

95050415, human unknown/unspecified, Lymphoblastoid

Synonym(s):

H929 Cell Line

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

NCI-H929, 95050415

biological source

human unknown/unspecified

growth mode

Suspension

karyotype

Near-tetraploid (8q+)

morphology

Lymphoblastoid

products

Immunoglobulin A (IgA)

receptors

Transferrin, CD38

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Human Caucasian IgA-producing plasmacytoma

Cell Line Description

This IgA-producing cell line has been established from a malignant effusion in a 62-year-old Caucasian woman with myeloma. They are positive for plasma cell antigen 1 (PCA-1), transferrin receptor and CD38 but negative for early B cell developmental markers. HLA DR and CALLA. NCI-H929 is negative for Epstein-Barr virus nuclear antigen. A rearrangement of the c-myc proto-oncogene has been observed as well as the expression of c-myc RNA and an activated ras allele.

Application

Chemotherapeutic agent studies, receptor studies, carcinogenesis assays.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11
D13S317: 12
D16S539: 9,13
D5S818: 11,12
D7S820: 10,12
THO1: 9.3
TPOX: 8,11
vWA: 14,15

Culture Medium

RPMI 1640 + 2mM Glutamine + 0.05mM 2-Mercaptoethanol (2ME) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 2-4 x 100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service