Skip to Content
MilliporeSigma
All Photos(1)

Documents

AT-3.1

94101451, rat skin, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

AT-3.1, 94101451

biological source

rat skin

growth mode

Adherent

karyotype

Hyperaneuploidy

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Rat dorsal prostatic adenocarcinoma

Cell Line Description

AT-3.1 is a subclone of the AT-3 cell line being one of a series of related variants originating from a single spontaneous adenocarcinoma in the dorsal lobe of the prostate of an aged male inbred Copenhagen rat. AT-3.1 was derived from an anaplastic tumour with a high rate of metastases and is androgen-independent.

Application

Carcinogenesis studies, cytogenetic analysis.

Culture Medium

RPMI 1640 + 2mM Glutamine + 250nM Dexamethasone (DXMT) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:10 to 1:20 i.e. seeding at 2-4 x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service