Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8D12

Reactivity: Anti-Guinea Pig Major Basic Protein, 94091320

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Mouse Hybridoma

ImmunoDon

Mouse spleen

Myeloma

Sp2/0

Reactivity

Anti-Guinea Pig Major Basic Protein

technique(s)

cell culture | mammalian: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse Sp2/0 x Mouse spleen

Cell Line Description

8D12 and 8A12 used in a sandwich ELISA giving a sensitive assay for biological samples. The antibody is directed against the guinea pig major basic protein found in guinea pig eosinophils. It can be paired in a sandwich ELISA with 8A12.

Immunogen

Guinea pig major basic protein

Application

Bioassay

Culture Medium

IDMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 1-2x100,000 cells/ml; 5% CO2; 37°C. Saturation density is reached at 1x1000,000 cells/ml.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service