Skip to Content
MilliporeSigma
All Photos(1)

Documents

JTC-27

94042255, rat liver, Epithelial-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

JTC-27, 94042255

biological source

rat liver

growth mode

Adherent

karyotype

Modal No52

morphology

Epithelial-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat hepatoma

Cell Line Description

Derived from a rat ascites hepatoma.

Application

Drug metabolism assays, hepatocyte studies, toxicological studies.

Culture Medium

DM-160 AU + 2mM Glutamine + 2% Foetal Bovine Serum (FBS) (10% for initial cell attachment after thawing).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:8 i.e. seeding at 1x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service