Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

537-B1-1

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 93102701, Reactivity: Anti-Lymphocytes/endothelium, sheep

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Mouse Hybridoma

packaging

tube of 5 μg 93102701-DNA-5UG
pkg of vial of cells 93102701-1VL

ImmunoDon

BALB/c mouse spleen

Myeloma

Sp2/O

Reactivity

Anti-Lymphocytes/endothelium, sheep

technique(s)

cell culture | mammalian: suitable

isotype

IgM

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse Sp2/O x Mouse BALB/c spleen

Cell Line Description

The antibody is directed against antigen expressed on most sheep lymphocytes and also endothelium. The antigen recognised by 537-B1-1 is down regulated after T cell activation.

Immunogen

Sheep lymphocytes

Culture Medium

IDMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS) (may benefit from IL-6 in medium)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service