Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

FW14.14.18

Reactivity: Anti-α-6 Integrin, human, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 93100512

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Unknown

packaging

tube of 5 μg 93100512-DNA-5UG
pkg of vial of cells 93100512-1VL

ImmunoDon

Not specified

Myeloma

Not specified

Reactivity

Anti-α-6 Integrin, human

technique(s)

cell culture | mammalian: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−196°C

Cell Line Description

The antibody is directed against human α-6 integrin and will block the binding of other α-6 integrin antibodies such as EA-1. The antibody specifically stains α-6 transfected cells.

Culture Medium

IDMEM + 2mM glutamine + 1% Hypoxanthine, Aminopterin, Thymidine (HAT) + 10% Foetal Bovine Serum (FBS) (may benefit from IL-6 in medium)

Subculture Routine

Maintain cultures between 3-5x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service