Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCL 4.1/F7

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 93031204, rat nerve (sheath), Flattened or crescent shaped

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

rat nerve (sheath)

packaging

tube of 5 μg 93031204-DNA-5UG
pkg of vial of cells 93031204-1VL

growth mode

Adherent

karyotype

42,XY / 43,XY+7

morphology

Flattened or crescent shaped

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat Schwann cell

Cell Line Description

This cell line was derived from neonatal Wistar rat Schwann cells being cultured for several months with intermittent exposure to the mitogen, cholera toxin and infrequent passaging to avoid premature transformation. After cloning one of these cultures by limiting dilution, the line SCL4.1/F7 was established growing without mitrogen. The cells display a subratum adhesion which is calcium-dependent in the millimolar range and pronounced contact-inhibition of growth. Confluent or subconfluent cultures readily cease proliferation and change to a differentiated (stellate / bipolar) morphology secreting an autocrine antiproliferative factor. Cells have been eradicated from mycoplasma at ECACC. With increasing passage cells have been shown to undergo transformation. It is recommended to growth-arrest cells for a few days before subculture to avoid rapid transformation.

Culture Medium

Ham′s F12 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split semi-confluent cultures 1:4 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C Saturation density is 10,000 cells/cm2.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service