Skip to Content
MilliporeSigma
All Photos(1)

Documents

S2.1

Reactivity: Anti-Swine Testis (ST) cell antigen, 92040116

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Pig x Pig Hybridoma

ImmunoDon

Boar spleen (d/d haplotype)

Myeloma

L231 (lymphoblastoid)

Reactivity

Anti-Swine Testis (ST) cell antigen

technique(s)

cell culture | mammalian: suitable

isotype

IgA

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Pig L231 x boar spleen (d/d haplotype)

Cell Line Description

The antibody is directed against the ST cell antigen as determined in virus enzyme linked cell immunoassays (VELCIA). The antibody does not cross-react with mouse 3T3 cells.

Immunogen

Swine testis cell culture supernatant containing transmissible gastro-enteritis virus (strain Nouzilly 188.SG)

Application

VELICA, ELISA

Culture Medium

RPMI 1640 + 2mM glutamine + 1mM Sodium Pyruvate (NaP) + 0.05mM 2-mercaptoethanol (2ME) + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 2-10x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service