Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F12

Reactivity: Anti-Haemolysin, Aeromonas species, 92032412

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Mouse Hybridoma

ImmunoDon

BALB/c mouse spleen

Myeloma

Sp2/0

Reactivity

Anti-Haemolysin, Aeromonas species

technique(s)

cell culture | mammalian: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse Sp2/0 x Mouse BALB/c spleen

Cell Line Description

The neutralising antibody is directed primarily against the haemolytic activity of Aeromanes sobria but it also neutralises cytotoxic activity. It recognises 3 major polypeptide bands of 52kDa, 47kDa, and 43kDa.

Immunogen

Aeromonas sobria haemidysin, partially purified 60kDa

Culture Medium

RPMI 1640 + 2mM glutamine + 1% Hypoxanthine, Aminopterin, Thymidine (HAT) + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service