Documents
MS
91070510, African green monkey kidney, Epithelial
About This Item
UNSPSC Code:
41106514
product name
MS, 91070510, from African green monkey kidney, Convention on the International Trade in Endangered Species of Wild Fauna and Flora (CITES)
biological source
African green monkey kidney
growth mode
Adherent
karyotype
Not specified
morphology
Epithelial
receptors
Not specified
relevant disease(s)
cancer
Cell Line Origin
Monkey African Green kidney
Cell Line Description
Derived from African green monkey. Useful for supporting functional co-cultures of rat hepatocytes. Originally mycoplasma infected.
Culture Medium
DMEM (D6546) + 2 mM L-Glutamine (G7513) + 5-10% FBS / FCS (F2442).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:3 to 1:6, i.e., seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37 °C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.