Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L52

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

Porcine blood

packaging

tube of 5 μg 91012321-DNA-5UG
pkg of vial of cells 91012321-1VL

growth mode

Suspension

karyotype

average No 36.5

morphology

Not specified

products

Immunoglobulin M (IgM)

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Porcine peripheral blood B cell

Cell Line Description

One of a series of cell lines established from mononuclear cells of a boar (Sus scrofa domestica L) which was injected with Shimozuma cells producing porcine retrovirus (Tsukuba-1). The cells contain retrovirus particles and are positive in RT assay.

DNA Profile

Not specified

Culture Medium

RPMI 1640 + 2 mM Glutamine + 1 mM Sodium Pyruvate (NaP) + 0.05 mM 2-Mercaptoethanol (2ME) + 10% Fetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 100,000 to 1,000,000 cells/ml; 5% CO2; 37 °C. Cells grow as small clumps.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service