Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

4647

90091902, vervet monkey kidney, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

4647, from vervet monkey kidney, 90091902, Convention on the International Trade in Endangered Species of Wild Fauna and Flora (CITES)

biological source

vervet monkey kidney

growth mode

Adherent

karyotype

Heterodiploid

morphology

Fibroblast

receptors

Not specified

Looking for similar products? Visit Product Comparison Guide

Cell Line Origin

Monkey Vervet kidney

Cell Line Description

Derived from kidney of a vervet monkey. Distribution restricted to non-profit organisations with involvement in virus vaccine research and development.

Culture Medium

DMEM (D6546) + 2 mM L-Glutamine (G7513) + 10% FBS / FCS (F2442).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6, i.e., seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37 °C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service