Skip to Content
MilliporeSigma
All Photos(1)

Documents

LLC-WRC 256

87061101, rat unknown/unspecified, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

LLC-WRC 256, 87061101, from rat unknown/unspecified

biological source

rat unknown/unspecified

growth mode

Adherent

karyotype

Modal no. 41 (2n=42)

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat Walker carcinoma epithelial

Cell Line Description

Walker rat tumour cell line, which supports the growth of herpes simplex, pseudorabies and vaccinia viruses. The cells are highly tumourigenic in suckling rats.

Culture Medium

Medium 199 + 2mM Glutamine + 5% Horse Serum (HS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2;37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service