Skip to Content
MilliporeSigma
All Photos(1)

Documents

SW 620 Cell Line human

87051203, human colon, Epithelial

Synonym(s):

SW620 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

SW 620 Cell Line human, 87051203

biological source

human colon

description

Human Caucasian colon adenocarcinoma

growth mode

Adherent

karyotype

2n = 46, hyperdiploid, modal no. 50

morphology

Epithelial

products

Carcinoembryonic antigen (CEA)

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Human Caucasian colon adenocarcinoma

Cell Line Description

Established from the lymph node of a 51 year old Caucasian male. The cells synthesise small quantities of CEA and are highly tumorigenic in nude mice. The established cell line consists of small spherical and bipolar cells resembling microvilli.The Y chromosome could not be detected in this cell line by short tandem repeat (STR)-PCR analysis when tested at ECACC. It is a known phenomenon that due to the increased genetic instability of cancer cell lines the Y chromosome can be rearranged or lost resulting in lack of detection. The cell line is identical to the source provided by the depositor based on the STR-PCR analysis.

Application

SW 620 cell line has been used:
  • as an in vitro model to study key cellular processes required for metastasis. It has also been used to study the effects of transforming growth factor-β (TGF-β) pathway and heat shock protein 90 (Hsp90) on the growth of cells
  • to study the relationship between zinc intake and risk of colon cancer
  • to test for intrahepatic tumor cell growth after intraportal injection
Tumourigenicity studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 13,14
D13S317: 12
D16S539: 9,13
D5S818: 13
D7S820: 8,9
THO1: 8
TPOX: 11
vWA: 16

Culture Medium

L15 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; No CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service