Skip to Content
MilliporeSigma
All Photos(1)

Documents

MRC-5 SV2

84100401, human lung (fetal), Fibroblast

Synonym(s):

MRC-5V2 Cells, MRC5V2 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

MRC-5 SV2, 84100401

biological source

human lung (fetal)

growth mode

Adherent

karyotype

Modal no. = 60-80, hyperdiploid

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Human foetal lung, SV40 transformed

Cell Line Description

Derived from the parental cell line MRC-5 by transformation with the SV40 virus. The cells do not senesce.

Application

Transformation and virus studies: SV40

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 11,12
D13S317: 11,14
D16S539: 11
D5S818: 11,12
D7S820: 10
THO1: 8
TPOX: 8
vWA: 15

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:5 to 1:10 i.e. seeding at 1-2x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service